
o-Tolylmagnesium chloride solution

Synonym(s):
2-Methylphenylmagnesium chloride
Linear Formula:
CH3C6H4MgCl
CAS Number:
Molecular Weight:
150.89
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing