Skip to Content
Merck
Octanoyl coenzyme A lithium salt hydrate ≥95% (HPLC)

Octanoyl coenzyme A lithium salt hydrate

Octanoyl coenzyme A lithium salt hydrate ≥95% (HPLC)
Synonym(s):
Capryloyl coenzyme A lithium salt hydrate
Empirical Formula (Hill Notation):
C29H50N7O17P3S · xLi+ · yH2O
CAS Number:
Molecular Weight:
893.73 (free acid basis)

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing