
Nebivolol Related Compound B

Synonym(s):
1-(6-Fluorochroman-2-yl)-2-{[2-(6-fluorochroman-2-yl)-2-hydroxyethyl]amino}ethanol hydrochloride, mixture of isomers
Empirical Formula (Hill Notation):
C22H25F2NO4 · HCl
Molecular Weight:
441.90
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing