Skip to Content
Merck
Myristic acid Pharmaceutical Secondary Standard; Certified Reference Material

Myristic acid

Myristic acid Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
1-Tridecanecarboxylic acid, C14:0, NSC 5028, Tetradecanoic acid, Myristic acid
Linear Formula:
CH3(CH2)12COOH
CAS Number:
Molecular Weight:
228.37
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Tissue
Quality Segment
Functional Group
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique