Monoclonal Anti-Phosphothreonine antibody produced in mouse
Synonym(s):
Monoclonal Anti-Phosphothreonine
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing