
Mogroside V

Empirical Formula (Hill Notation):
C60H102O29
CAS Number:
Molecular Weight:
1287.43
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing