Skip to Content
Merck
Metolachlor PESTANAL®, analytical standard

Metolachlor

Metolachlor PESTANAL®, analytical standard
Synonym(s):
2-Chloro-N-(2-ethyl-6-methylphenyl)-N-(-2-methoxy-1-methylethyl)acetamide
Empirical Formula (Hill Notation):
C15H22ClNO2
CAS Number:
Molecular Weight:
283.79
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing