Magnolol
Synonym(s):
2,2′-Bichavicol, 5,5′-Diallyl-2,2′-biphenyldiol
Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Activity Spectrum
Format
Research Area
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing