
Luteinizing hormone releasing hormone human acetate salt

Synonym(s):
Gonadoliberin, Gonadotropin releasing hormone, LH-RH
Empirical Formula (Hill Notation):
C55H75N17O13
CAS Number:
Molecular Weight:
1182.29
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing