
Loratadine for system suitability

Synonym(s):
Loratadine, 4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene-1-piperidinecarboxylic acid ethyl ester, Loratidine
Empirical Formula (Hill Notation):
C22H23N2O2Cl
CAS Number:
Molecular Weight:
382.88
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing