Skip to Content
Merck
Lanthanum(III) nitrate hexahydrate 99.99% trace metals basis

Lanthanum(III) nitrate hexahydrate

Lanthanum(III) nitrate hexahydrate 99.99% trace metals basis
Synonym(s):
Nitric acid, lanthanum (III) salt, hexahydrate
Linear Formula:
La(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
433.01
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-2 of 2