
Lamotrigine Related Compound B

Synonym(s):
2,3-Dichlorobenzoic acid
Linear Formula:
Cl2C6H3CO2H
CAS Number:
Molecular Weight:
191.01
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing