
Ibandronate sodium

Synonym(s):
Ibandronate monosodium salt monohydrate
Empirical Formula (Hill Notation):
C9H22NNaO7P2 · H2O
CAS Number:
Molecular Weight:
359.23
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2