
Hydrazine sulfate-15N2

Synonym(s):
15N Labeled hydrazine sulfate
Linear Formula:
H215N15NH2 · H2SO4
CAS Number:
Molecular Weight:
132.11
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing