
Glycopyrrolate Related Compound C

Synonym(s):
α-Cyclopentylmandelic acid, 2-Cyclopentyl-2-hydroxy-2-phenylacetic acid
Empirical Formula (Hill Notation):
C13H16O3
CAS Number:
Molecular Weight:
220.26
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2