
Fosfomycin tromethamine

Synonym(s):
Fosfomycin trometamol
Empirical Formula (Hill Notation):
C7H18NO7P
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing