
Fibrinopeptide A human

Empirical Formula (Hill Notation):
C63H97N19O26
CAS Number:
Molecular Weight:
1536.56
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing