Skip to Content
Merck
Ethyl methyl carbonate Technipur®, 99.9%, ≤10 ppm acid, ≤10 ppm H2O

Ethyl methyl carbonate

Ethyl methyl carbonate Technipur®, 99.9%, ≤10 ppm acid, ≤10 ppm H2O
Synonym(s):
Carbonic acid ethyl methyl ester, EMC, Methyl ethyl carbonate
Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Showing 1-4 of 4
Compare
Product Number
Description
Pricing