
Emetine dihydrochloride

Synonym(s):
6′,7′,10,11-Tetramethoxyemetan dihydrochloride, Emetine, Dihydrochloride
Empirical Formula (Hill Notation):
C29H40N2O4 · 2HCl
CAS Number:
Molecular Weight:
553.56
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Activity Spectrum
Format
Research Area
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Showing 1-3 of 3