
E7820-PEG3-NH2 hydrochloride

Empirical Formula (Hill Notation):
C25H31N5O6S · xHCl
Molecular Weight:
529.61 (free base basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Reactivity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing