Skip to Content
Merck
DMT-locMeC(bz) Phosphoramidite configured for PerkinElmer

DMT-locMeC(bz) Phosphoramidite

DMT-locMeC(bz) Phosphoramidite configured for PerkinElmer
Synonym(s):
DMT-locMeC(bz) Phosphoramidite
Empirical Formula (Hill Notation):
C48H54N5O9P
CAS Number:
Molecular Weight:
875.94

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-3 of 3