
Digoxin for peak identification

Synonym(s):
Digoxin, 12β-Hydroxydigitoxin
Empirical Formula (Hill Notation):
C41H64O14
CAS Number:
Molecular Weight:
780.94
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing