
Darifenacin hydrobromide

Synonym(s):
(3S)-1-[2-(2,3-Dihydro-5-benzofuranyl)ethyl]-α,α-diphenyl-3-pyrrolidineacetamide hydrobromide, UK 88525-04
Empirical Formula (Hill Notation):
C28H30N2O2 · HBr
CAS Number:
Molecular Weight:
507.46
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Research Area
Quality Segment
Application
API Family
Showing 1-2 of 2
Compare
Product Number
Description
Pricing