Skip to Content
Merck
Calcium lactobionate monohydrate 98.0-102.0% (EDTA titration)

Calcium lactobionate monohydrate

Calcium lactobionate monohydrate 98.0-102.0% (EDTA titration)
Synonym(s):
4-O-β-D-Galactopyranosyl-D-gluconic acid hemicalcium salt monohydrate
Empirical Formula (Hill Notation):
C12H21O12 · 0.5Ca · H2O
CAS Number:
Molecular Weight:
395.34
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing