
Bifenthrin

Synonym(s):
3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic acid (2-methylbiphenyl-3-yl)methyl ester
Empirical Formula (Hill Notation):
C23H22ClF3O2
CAS Number:
Molecular Weight:
422.87
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing