Skip to Content
Merck
Berberine chloride British Pharmacopoeia (BP) Reference Standard

Berberine chloride

Berberine chloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Berberine chloride form, Natural Yellow 18
Empirical Formula (Hill Notation):
C20H18ClNO4
CAS Number:
Molecular Weight:
371.81

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique