
Bacteria stain propidium iodide solution

Synonym(s):
Propidium iodide solution, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide, 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium, diiodide
Empirical Formula (Hill Notation):
C27H34I2N4
CAS Number:
Molecular Weight:
668.39
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing