Skip to Content
Merck
Angiotensin I human acetate salt hydrate ≥90% (HPLC), powder, suitable for mammalian cell culture

Angiotensin I human acetate salt hydrate

Angiotensin I human acetate salt hydrate ≥90% (HPLC), powder, suitable for mammalian cell culture
Synonym(s):
Hypertensin I
Empirical Formula (Hill Notation):
C62H89N17O14 · xC2H4O2 · yH2O
CAS Number:
Molecular Weight:
1296.48 (anhydrous free base basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1