
Amrinone

Synonym(s):
5-Amino-(3,4′-bipyridin)-6(1H)-one, Inamrinone, Amrinone
Empirical Formula (Hill Notation):
C10H9N3O
CAS Number:
Molecular Weight:
187.20
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing