
4-Amino-1-naphthalenesulfonic acid sodium salt hydrate

Synonym(s):
1-Naphthylamine-4-sulfonic acid sodium salt, Naphthionic acid sodium salt, Sodium 4-amino-1-naphthalenesulfonate
Linear Formula:
H2NC10H6SO3Na · xH2O
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing