
3-(N-Ethyl-3-methylanilino)propanesulfonic acid sodium salt

Synonym(s):
3-(N-Ethyl-m-toluidino)propanesulfonic acid sodium salt, 3-(Ethyl[3-methylphenyl]amino)-1-propanesulfonic acid, N-Ethyl-N-sulfopropyl-m-toluidine sodium salt, Sodium 3-(N-ethyl-3-methylanilino)propanesulfonate, Sodium N-ethyl-N-(3-sulfopropyl)-m-toluidine, TOPS
Empirical Formula (Hill Notation):
C12H18NNaO3S
CAS Number:
Molecular Weight:
279.33
Products
Product Category
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing