3-Methyl-3-oxetanemethanol
Synonym(s):
3-Hydroxymethyl-3-methyl-oxetane
Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1