Skip to Content
Merck
3-Fluoro-2-methoxyaniline 97%

3-Fluoro-2-methoxyaniline

3-Fluoro-2-methoxyaniline 97%
Synonym(s):
3-Fluoro-2-methoxybenzenamine, 3-Fluoro-2-methoxyphenylamine, 3-Fluoro-o-anisidine
Linear Formula:
FC6H3(OCH3)NH2
CAS Number:
Molecular Weight:
141.14

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing