
3,5,6-Trichloro-2-hydroxybenzoic acid

Synonym(s):
3,5,6-Trichlorosalicylic acid
Linear Formula:
HC6Cl3-2-(OH)CO2H
CAS Number:
Molecular Weight:
241.46
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing