Skip to Content
Merck
2-Keto-3-deoxyoctonate ammonium salt ≥97%

2-Keto-3-deoxyoctonate ammonium salt

2-Keto-3-deoxyoctonate ammonium salt ≥97%
Synonym(s):
3-Deoxyoctulosonic acid, KDO
Linear Formula:
C8H14O8 · NH3
CAS Number:
Molecular Weight:
255.22

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing