Skip to Content
Merck
2′-Deoxycytidine 5′-triphosphate disodium salt 100 mM (pH 7)

2′-Deoxycytidine 5′-triphosphate disodium salt

2′-Deoxycytidine 5′-triphosphate disodium salt 100 mM (pH 7)
Synonym(s):
dCTP-Na2
Empirical Formula (Hill Notation):
C9H14N3Na2O13P3
CAS Number:
Molecular Weight:
511.12

Products

Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2