Skip to Content
Merck
2-Butyl-4-chloro-5-(hydroxymethyl)imidazole 97%

2-Butyl-4-chloro-5-(hydroxymethyl)imidazole

2-Butyl-4-chloro-5-(hydroxymethyl)imidazole 97%
Empirical Formula (Hill Notation):
C8H13ClN2O
CAS Number:
Molecular Weight:
188.65

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing