Skip to Content
Merck
2,2-Dimethoxy-2-phenylacetophenone 99%

2,2-Dimethoxy-2-phenylacetophenone

2,2-Dimethoxy-2-phenylacetophenone 99%
Synonym(s):
α,α-Dimethoxy-α-phenylacetophenone, Benzil α,α-dimethyl acetal
Linear Formula:
C6H5COC(OCH3)2C6H5
CAS Number:
Molecular Weight:
256.30
EC Number:

Products

Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing