
1H,1H,2H,2H-Perfluorooctyltriethoxysilane

Synonym(s):
Triethoxy(1H,1H,2H,2H-perfluoro-1-octyl)silane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octyl)silane
Empirical Formula (Hill Notation):
C14H19F13O3Si
Products
Product Category
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing