
1,3,4,6-Tetrachloro-3α,6α-diphenylglycouril

Synonym(s):
Iodo-Gen®, NSC 4462
Empirical Formula (Hill Notation):
C16H10Cl4N4O2
CAS Number:
Molecular Weight:
432.09
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing