Skip to Content
Merck
(1,2-Methanofullerene C60)-61-carboxylic acid 97% (HPLC)

(1,2-Methanofullerene C60)-61-carboxylic acid

(1,2-Methanofullerene C60)-61-carboxylic acid 97% (HPLC)
Empirical Formula (Hill Notation):
C62H2O2
CAS Number:
Molecular Weight:
778.68

Products

Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing