Skip to Content
Merck
All Photos(1)

Key Documents

1686386

USP

Triglycine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Gly-Gly-Gly, Glycyl-glycyl-glycine, Triglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH2CONHCH2COOH
CAS Number:
Molecular Weight:
189.17
Beilstein:
1711130
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triglycine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NCC(=O)NCC(=O)NCC(O)=O

InChI

1S/C6H11N3O4/c7-1-4(10)8-2-5(11)9-3-6(12)13/h1-3,7H2,(H,8,10)(H,9,11)(H,12,13)

InChI key

XKUKSGPZAADMRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Gly-Gly

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alosetron Hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Also, for use with USP monograph such as Alosetron Tablets

Biochem/physiol Actions

Substrate for reproducible serum protein measurements by the biuret reaction and for the assay of aminotripeptidases.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycine
United States Pharmacopeia, 45(1), 2134-2134 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service