Skip to Content
Merck
All Photos(8)

Key Documents

ZMS1212

Sigma-Aldrich

Anti-p-Ser/Thr-Pro MPM-2 Antibody, clone MPM-2 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-NKX2-1 Antibody, clone 1H5-H2 ZooMAb® Rabbit Monoclonal, phospho-MPM2 (Ser/Thr)-Pro,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MPM-2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt Variable kDa
observed mol wt ~50-180 kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone MPM-2 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects phosphorylated serines/threonines when followed by a proline in mitotic proteins.

Immunogen

Mitotic HeLa cell cytosolic lysate.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Human skin tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected phospho-S/T-Pro MPM-2 in Human skin tissue sections.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected phospho-S/T-Pro MPM-2 in Human prostate cancer tissue sections

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected phospho-S/T-Pro MPM-2 in HeLa cells.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho-S/T-Pro MPM-2 in HeLa cells treated with colcemid.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Upon entry into Mitotic (M) phase of cell cycle, many proteins are phosphorylated either directly or indirectly by M-phase-promoting factor (MPF). The reversible phosphorylation of proteins regulates transitions between various phases of the cell cycle and the cytoarchitectural changes necessary for cell division. One such group of proteins is the family of MAP kinases that play critical roles in cell cycle control. They mediate intracellular signal transmission in response to external stimuli. p42MAP kinase requires phosphorylation on both threonine 183 and tyrosine 185 for its full activation. This MPM-2 recombinant monoclonal antibody binds to a phospho amino acid-containing epitopes present on several proteins of M-phase eukaryotic cells. It recognizes phosphorylated serine/threonine epitope that is followed by a proline residue. It is reported to recognizes the activated form of p42 MAP kinase. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, o

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service