Skip to Content
Merck
All Photos(1)

Key Documents

SML1366

Sigma-Aldrich

CVT-10216

≥98% (HPLC)

Synonym(s):

3-[[[3-[4-[(Methylsulfonyl)amino]phenyl]-4-oxo-4H-1-benzopyran-7-yl]oxy]methyl]-benzoic acid, 3-[[[3-[4-[(Methylsulfonyl)amino]phenyl]-4-oxo-4H-chromen-7-yl]oxy]methyl]benzoic acid, CVT 10216, GS 455534

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H19NO7S
CAS Number:
Molecular Weight:
465.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)C1=CC(COC2=CC=C3C(OC=C(C4=CC=C(NS(C)(=O)=O)C=C4)C3=O)=C2)=CC=C1

InChI

1S/C24H19NO7S/c1-33(29,30)25-18-7-5-16(6-8-18)21-14-32-22-12-19(9-10-20(22)23(21)26)31-13-15-3-2-4-17(11-15)24(27)28/h2-12,14,25H,13H2,1H3,(H,27,28)

InChI key

YYOOFJZTRCPVFD-UHFFFAOYSA-N

General description

CVT-10216 is a 5-HT2c agonist.

Biochem/physiol Actions

CVT-10216 is a selective reversible inhibitor of aldehyde dehydrogenase-2 (ALDH-2), thereby inhibiting dopamine synthesis. CVT-10216 is active in suppressing alcohol and cocaine use, and anxiety.
CVT-10216 possesses unequivocal anxiolytic properties. It will not affect the suppression of locomotor activity by mCPP (m-chlorophenylpiperazine).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Yuan et al.
PloS one, 15(11), e0242809-e0242809 (2020-12-01)
The bone marrow microenvironment (BME) in acute myeloid leukemia (AML) consists of various cell types that support the growth of AML cells and protect them from chemotherapy. Mesenchymal stromal cells (MSCs) in the BME have been shown to contribute immensely
Simin Zhang et al.
Aesthetic plastic surgery, 44(3), 993-1005 (2020-01-19)
The cutaneous wound healing process mainly comprises re-epithelialization, fibrosis, and neovascularization. Impaired wound healing is common but tricky in plastic surgery. Aldehyde dehydrogenase 2 (ALDH2), the most effective subset of the ALDH enzyme family, is known to exert a major
David H Overstreet et al.
Pharmacology, biochemistry, and behavior, 94(2), 255-261 (2009-09-15)
CVT-10216 is a highly selective, reversible inhibitor of ALDH-2 that reduces excessive alcohol drinking. Anxiety plays a role in alcoholism. The present study asks whether CVT-10216 has anxiolytic properties, as reflected in social interaction behavior in four unrelated rodent models:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service