Skip to Content
Merck
All Photos(2)

Key Documents

RAB0493

Sigma-Aldrich

Mouse Osteoprotegerin ELISA Kit

for serum, plasma, cell culture supernatant

Synonym(s):

OPG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 1.65-1200 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The Mouse OPG (Osteoprotegerin) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse OPG in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse Osteoprotegerin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Simpson et al.
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA, 25(10), 2383-2388 (2014-06-15)
We compared circulating levels of Wnt inhibitors among patients with high bone mass mutations in LRP5, unaffected kindred, and unrelated normal controls. Inhibitors were unchanged in affected and unaffected kindred. We saw no meaningful differences between controls and affected individuals.
Stella Bernardi et al.
Molecular and cellular endocrinology, 394(1-2), 13-20 (2014-07-08)
Inflammation is believed to link obesity to insulin resistance, as in the setting of metabolic syndrome (MetS). Osteoprotegerin (OPG) is a soluble protein that seems to exert proatherogenic and prodiabetogenic effects. This study aims at determining OPG levels in MetS
Regulation of Osteoblast Differentiation by Acid-Etched and/or Grit-Blasted Titanium Substrate Topography Is Enhanced by 1,25(OH)2D3 in a Sex-Dependent Manner.
Olivares-Navarrete R, Hyzy SL, Boyan BD, et al.
BioMed Research International, 2015, 365014-365014 (2015)
Gen Kuroyanagi et al.
Prostaglandins, leukotrienes, and essential fatty acids, 91(3), 73-80 (2014-05-13)
Resveratrol, a natural polyphenol with health-related properties mainly existing in grape skins and red wine, possesses beneficial effects on human being. We have previously reported that prostaglandin D2 (PGD2) stimulates heat shock protein 27 (HSP27) induction via activation of p44/p42
Jonggun Kim et al.
Journal of functional foods, 8, 367-376 (2014-07-19)
The inverse relationship between osteoblast and adipocyte differentiation in bone marrow mesenchymal stem cells has been linked to overall bone mass. It has previously been reported that conjugated linoleic acid (CLA) inhibits adipogenesis via a peroxisome-proliferator activated receptor-γ (PPARγ) mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service