Key Documents
BMSIPS205040E
Sigma-Aldrich
Benchmark OS-40L Overhead Stirrer
AC/DC input 220 V, Schuko plug
Synonym(s):
masher, bag homogeniser, bag blender
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-40-E
parameter
30-2200 rpm speed
General description
Benchmark′s Overhead Stirrers feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples. Speed and run time are accurately set and monitored on the LCD screen. They are supplied with a 4 arm stainless steel propeller, additional steel and PTFE accessories are available. A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools. The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction, while a splash cover protects the connection mechanism from potential corrosion. Unlike most mixers, the Benchmark series is also supplied with a support stand and mounting hardware. The stirrer easily and securely attaches to the stand for compatibility with a variety of vessels, up to 24 inches high.
Features and Benefits
LCD control panelIncludes complete stand assemblyCompact and quietDigital accuracy and convenience