Skip to Content
Merck
All Photos(1)

Key Documents

20966

Sigma-Aldrich

Cesium chloride

BioUltra, for molecular biology, ≥99.5% (AT)

Synonym(s):

CsCl, cesium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

product line

BioUltra

Assay

≥99.5% (AT)

form

solid

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.001% total nitrogen (N)

loss

≤0.1% loss on drying, 110 °C

pH

6.0-7.5 (25 °C, 3 M in H2O)

mp

645 °C (lit.)

solubility

H2O: 3 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤20 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

3 M in H2O

UV absorption

λ: 260 nm Amax: 0.02
λ: 280 nm Amax: 0.02

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cesium chloride (CsCl) can be used for the preparation of electrically conducting glasses. It can also be used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Other Notes

Cesium chloride gradient centrifugation for phage purification; Isolation of satellite DNA; RNA purification; Buoyant density of DNA - Hoechst 33258 complexes in CsCl gradients.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Karlovsky et al.
Analytical biochemistry, 194(1), 192-197 (1991-04-01)
Buoyant density of DNA in CsCl gradients with Hoechst 33258 (bisbenzimide) was investigated as a function of guanine plus cytosine content of the DNA (%GC; in mole percent). A formula for calculating %GC from the refractive index (nD) of the
Turpen, T. H. and Griffith, O. M.
Biotechnology, 4, 11-11 (1986)
Cooney, C. A. et al.
Methods in Molecular Biology, 2, 21-21 (1984)
Miller, H.
Methods in Enzymology, 152, 159-159 (1987)
Borja Muñoz-Solano et al.
Toxins, 12(6) (2020-06-11)
Mycotoxins are toxic compounds for humans and animals that are produced by fungi. Mycotoxin contamination in feed is a global safety concern and effective control of these compounds in this matrix is needed. This study proposes a simple, cost-effective analytical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service