Skip to Content
Merck
All Photos(1)

Key Documents

NIST8323

Yeast Protein Extract

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

solution

packaging

pkg of 3 vials x 0.2 mL

manufacturer/tradename

NIST®

application(s)

food and beverages

storage temp.

−70°C

General description

Reference Material (RM) 8323 is intended to support measurements used to identify proteins in complex protein mixtures such as those used in proteomics. RM 8323 can be used to help assess measurement repeatability within a laboratory or comparability between laboratories or among different measurement approaches [1]. RM 8323 can also be used in the development and validation of new measurement approaches for identifying proteins in complex protein mixtures. A unit of RM 8323 consists of three vials, each containing 200μL of frozen yeast protein extract solution.The proteins extracted from Saccharomyces cerevisiaehave been solubilized in 50 mmol/L ammonium bicarbonate in water.

SRM 8323_cert

SRM 8323_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Protein

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service