Skip to Content
Merck
All Photos(1)

Key Documents

90058

Millipore

Inulin Discs

suitable for microbiology, Sterile filter paper discs impregnated with inulin

Synonym(s):

Carbohydrate discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

Quality Level

sterility

sterile

product line

BioChemika

form

disc

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 10 × 25 discs

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Streptococcus spp.
bacteria

General description

Inulin Discs are used to differentiate bacteria based on the inulin fermentation ability. Microorganisms can only ferment certain carbohydrates, depending on the enzymes they possess to break down these carbohydrates. Fermentation can be detected by gas production (CO2) in liquid media and/or color change of pH caused by acid production. The inulin discs are aseptically added to any sugar-free basal media to test the ability of the organism to hydrolyze inulin, a naturally occurring polysaccharide, as the sole source of carbon and energy during fermentation. Inulin is typically fermented under acid production by many Streptococcus species.Inulin is used for the differentiation of streptococci, for example, to differentiate Streptococcus pneumoniae from other non-pathogenic Streptococcus species. According to CDC, inulin is also used for the identification of Globicatella species.

Application

Inulin Discs are recommended to detect inulin fermenting bacteria in food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nopporn Jongkamonwiwat et al.
Cell reports, 33(8), 108431-108431 (2020-11-26)
Exposure to excessive sound causes noise-induced hearing loss through complex mechanisms and represents a common and unmet neurological condition. We investigate how noise insults affect the cochlea with proteomics and functional assays. Quantitative proteomics reveals that exposure to loud noise
Martin Golkowski et al.
Cell systems, 11(2), 196-207 (2020-08-07)
Hepatocellular carcinoma (HCC) is a complex and deadly disease lacking druggable genetic mutations. The limited efficacy of systemic treatments for advanced HCC implies that predictive biomarkers and drug targets are urgently needed. Most HCC drugs target protein kinases, indicating that
Lior Soday et al.
Cell reports, 27(6), 1920-1933 (2019-05-09)
Vaccinia virus (VACV) has numerous immune evasion strategies, including multiple mechanisms of inhibition of interferon regulatory factor 3 (IRF-3), nuclear factor κB (NF-κB), and type I interferon (IFN) signaling. Here, we use highly multiplexed proteomics to quantify ∼9,000 cellular proteins and
Heidi M Blank et al.
Molecular biology of the cell, 31(10), 1069-1084 (2020-03-05)
Establishing the pattern of abundance of molecules of interest during cell division has been a long-standing goal of cell cycle studies. Here, for the first time in any system, we present experiment-matched datasets of the levels of RNAs, proteins, metabolites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service