Skip to Content
Merck
All Photos(1)

Key Documents

74239

Supelco

Tetraconazole

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(RS)-2-(2,4-Dichlorophenyl)-3-(1H-1,2,4-triazolyl)propyl 1,1,2,2-tetrafluoroethyl ether, (±)-1-[2-(2,4-Dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]-1H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11Cl2F4N3O
CAS Number:
Molecular Weight:
372.15
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

100
300

product line

TraceCERT®

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

FC(F)C(F)(F)OCC(Cn1cncn1)c2ccc(Cl)cc2Cl

InChI

1S/C13H11Cl2F4N3O/c14-9-1-2-10(11(15)3-9)8(4-22-7-20-6-21-22)5-23-13(18,19)12(16)17/h1-3,6-8,12H,4-5H2

InChI key

LQDARGUHUSPFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Tetraconazole is a systemic triazole fungicide― known for its protective, curative, and eradicant activities. It inhibits ergosterol biosynthesis by interfering with the activity of C14-demethylase. It is used for protection against:
  • Puccinia spp. and Ustilago spp. in cereals
  • Eysiphe betae, Cercospora beticola and Uromyces betae in sugar beets
  • Podosphaera leucotricha and Venturia spp. in pome fruit
  • Erysiphe necator in grapes
  • Sphaerotheca fuliginea and Erysiphe cichoracearum in cucurbits
  • Sphaerotheca macularis in strawberries
  • Leveillula taurica in tomatoes, peppers, and artichokes
  • Powdery mildew, rust and scab in vegetables, ornamentals, and tropical fruit
According to the Commission Implementing Regulation (EU) No 540/2011 of 25th May 2011, tetraconazole is approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009. But maximum residue levels of 0.01 mg/kg to 1 mg/kg are allowed for the presence of tetraconazole in products of plant and animal origin, according to the EU Reg. 2019/1015.
Tetraconazole has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Tetraconazole CRM may also be used as given below:

  • Multi-residue analysis of 213 pesticides in edible oil samples using an analytical approach combining QuEChERS methodology with gas chromatography-tandem mass spectrometry (GC-MS/MS)
  • Multi-residue analysis of 379 pesticides in human serum samples extracted by QuEChERS method― using liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Detection of 327 multiclass pesticide and 17 veterinary drug residues in eight animal feed samples using liquid- and gas- chromatography combined with tandem mass spectrometry (LC-MS/MS and GC-MS/MS)
  • Development of a semi-automated method for the analysis of 261 pesticides and 41 environmental contaminants in catfish muscle samples by QuEChERS sample extraction and low-pressure GC-MS/MS and UHPLC-MS/MS, following an automated robotic micro-SPE clean-up
  • Multi-residue analysis of 67 pesticides in olive oil, olives, and avocado samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) to evaluate various clean-up sorbents
  • Method development and validation for the determination of 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high resolution tandem mass spectrometry

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service